4-Indanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydro-1H-inden-4-ol

074542-5G
102536-966EA 572.31 USD
102536-966
4-Indanol ≥95%
4-Indanol

Hazard: Irritant/Refrigerate

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00053431
CAS Number: 1641-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR