2,7-Diazaspiro[4.5]decan-1-one ≥95%

Supplier: Matrix Scientific

074539-1G
102536-950EA 992.31 USD
102536-950
2,7-Diazaspiro[4.5]decan-1-one ≥95%
2,7-Diazaspiro[4.5]decan-1-one

Hazard: Irritant

Formula: C₈H₁₄N₂O
CAS Number: 887118-43-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR