tert-Butyl-4-carbamothioylpiperazine-1-carboxylate ≥95%

Supplier: Matrix Scientific

074536-1G 074536-10G
102536-938EA 1978.46 USD
102536-938 102536-934
tert-Butyl-4-carbamothioylpiperazine-1-carboxylate ≥95%
tert-Butyl-4-carbamothioylpiperazine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₉N₃O₂S
MW: 245.34 g/mol
MDL Number: MFCD07787249
CAS Number: 196811-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR