2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide ≥95%

074535-250MG
102536-930EA 1397.37 USD
102536-930
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide ≥95%
2-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetamide

Hazard: Irritant

Formula: C₁₁H₁₈BN₃O₃
MW: 251.0899 g/mol
MDL Number: MFCD16660236
CAS Number: 1083326-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR