4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-propanenitrile ≥95%

Supplier: Matrix Scientific

074533-1G
102536-922EA 400 USD
102536-922
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-propanenitrile ≥95%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole-1-propanenitrile

Hazard: Irritant

Formula: C₁₂H₁₈BN₃O₂
MW: 247.1012 g/mol
MDL Number: MFCD16660233
CAS Number: 1022092-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR