3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride ≥95%

Supplier: Matrix Scientific

074531-1G
102536-914EA 946.15 USD
102536-914
3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride ≥95%
3-Oxa-8-azabicyclo[3.2.1]octane hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO
MW: 149.6186 g/mol
MDL Number: MFCD18432562
CAS Number: 904316-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR