4H-Cyclopenta[b]thiophen-6(5H)-one ≥95%

Supplier: Matrix Scientific

074530-250MG 074530-5G
102536-906EA 283.08 USD
102536-906 102536-910
4H-Cyclopenta[b]thiophen-6(5H)-one ≥95%
4H-Cyclopenta[b]thiophen-6(5H)-one

Hazard: Irritant

Formula: C₇H₆OS
MW: 138.1869 g/mol
MDL Number: MFCD03426936
CAS Number: 5650-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR