4-Hydroxycyclohexanecarbonitrile ≥95%

Supplier: Matrix Scientific

074528-1G
102536-898EA 584.62 USD
102536-898
4-Hydroxycyclohexanecarbonitrile ≥95%
4-Hydroxycyclohexanecarbonitrile

Hazard: Irritant

Formula: C₇H₁₁NO
MW: 125.1700 g/mol
Boiling Pt: 153…154 °C (12 Torr)
MDL Number: MFCD13688990
CAS Number: 24056-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR