2-Bromo-5-fluorobenzyl alcohol ≥95%
Supplier: Matrix Scientific
074527-100G
074527-5G
102536-894EA
880
USD
102536-894
102536-890
2-Bromo-5-fluorobenzyl alcohol ≥95%
2-Bromo-5-fluorobenzyl alcohol
Hazard: Irritant
Formula:
C₇H₆BrFO MW: 205.03 g/mol |
MDL Number:
MFCD00142876 CAS Number: 202865-66-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...