3-(Iodomethyl)-1,1-dimethoxycyclobutane ≥95%

Supplier: Matrix Scientific

074524-1G
102536-874EA 1383.08 USD
102536-874
3-(Iodomethyl)-1,1-dimethoxycyclobutane ≥95%
3-(Iodomethyl)-1,1-dimethoxycyclobutane

Hazard: Irritant

Formula: C₇H₁₃IO₂
CAS Number: 1003013-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR