Benzyl-4-hydroxyazepane-1-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Benzyl 4-hydroxyazepane-1-carboxylate
074523-5G
074523-250MG
102536-870EA
2336.92
USD
102536-870
102536-866
Benzyl-4-hydroxyazepane-1-carboxylate ≥95%
Benzyl-4-hydroxyazepane-1-carboxylate
Hazard: Irritant
Formula:
C₁₄H₁₉NO₃ MW: 249.3100 g/mol |
MDL Number:
MFCD07778596 CAS Number: 648418-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...