4-Bromoisoindolin-1-one ≥95%

Supplier: Matrix Scientific

074520-1G
102536-850EA 156.92 USD
102536-850
4-Bromoisoindolin-1-one ≥95%
4-Bromoisoindolin-1-one

Hazard: Irritant

Formula: C₈H₆BrNO
MW: 212.0400 g/mol
MDL Number: MFCD09701286
CAS Number: 337536-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR