Ethyl-3-methyl-1H-pyrrole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-methyl-1H-pyrrole-2-carboxylate

074519-1G
102536-844EA 75.38 USD
102536-844
Ethyl-3-methyl-1H-pyrrole-2-carboxylate ≥95%
Ethyl-3-methyl-1H-pyrrole-2-carboxylate

Hazard: Irritant

Formula: C₈H₁₁NO₂
MW: 153.1784 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD01006747
CAS Number: 3284-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR