4-Chloro-5-fluoro-2-methylpyrimidine ≥95%

Supplier: Matrix Scientific

074518-1G
102536-840EA 769.23 USD
102536-840
4-Chloro-5-fluoro-2-methylpyrimidine ≥95%
4-Chloro-5-fluoro-2-methylpyrimidine

Hazard: Irritant

Formula: C₅H₄ClFN₂
MW: 146.5501 g/mol
MDL Number: MFCD09834964
CAS Number: 898044-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR