tert-Butyl piperazin-1-ylcarbamate ≥95%

Supplier: Matrix Scientific

074516-1G
102536-828EA 983.08 USD
102536-828
tert-Butyl piperazin-1-ylcarbamate ≥95%
tert-Butyl piperazin-1-ylcarbamate

Hazard: Irritant

Formula: C₉H₁₉N₃O₂
MW: 201.2661 g/mol
MDL Number: MFCD11840387
CAS Number: 147081-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR