Methyl-6-aminopicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Aminopicolinic acid methyll ester6-Aminopyridine-2-carboxylic acid methyll esterMethyl 6-AminopicolinateMethyl 6-Aminopyridine-2-carboxylateMethyl 6-aminopicolinate

074515-5G
102536-822EA 44.62 USD
102536-822
Methyl-6-aminopicolinate ≥95%
Methyl 6-aminopicolinate

Hazard: Irritant

Formula: C₇H₈N₂O₂
MW: 152.1500 g/mol
MDL Number: MFCD00233712
CAS Number: 36052-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR