5-Bromo-4-methylpyridine-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074511-5G
102536-800EA 292.31 USD
102536-800
5-Bromo-4-methylpyridine-2-carboxylic acid ≥95%
5-Bromo-4-methylpyridine-2-carboxylic acid

Hazard: Irritant

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
MDL Number: MFCD07375100
CAS Number: 886365-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR