4-(Dibenzylamino)cyclohexanone ≥95%

Supplier: Matrix Scientific

074507-1G
102536-778EA 144.62 USD
102536-778
4-(Dibenzylamino)cyclohexanone ≥95%
4-(Dibenzylamino)cyclohexanone

Hazard: Irritant

Formula: C₂₀H₂₃NO
MW: 293.4000 g/mol
MDL Number: MFCD16659682
CAS Number: 149506-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR