3,4-Bis(trifluoromethyl)phenylboronic acid ≥95%

Supplier: Matrix Scientific

074503-5G
102536-756EA 1383.08 USD
102536-756
3,4-Bis(trifluoromethyl)phenylboronic acid ≥95%
3,4-Bis(trifluoromethyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₅BF₆O₂
CAS Number: 1204745-88-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR