2-Chloro-3-(chloromethyl)pyrazine ≥95%

Supplier: Matrix Scientific

074501-250MG 074501-5G
102536-744EA 640 USD
102536-744 102536-748
2-Chloro-3-(chloromethyl)pyrazine ≥95%
2-Chloro-3-(chloromethyl)pyrazine

Hazard: Irritant

Formula: C₅H₄Cl₂N₂
CAS Number: 45660-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR