2-Bromo-5-fluoro-3-methylpyridine ≥95%

074500-5G
102536-740EA 2127.27 USD
102536-740
2-Bromo-5-fluoro-3-methylpyridine ≥95%
2-Bromo-5-fluoro-3-methylpyridine

Hazard: Irritant

Formula: C₆H₅BrFN
MW: 190.01 g/mol
MDL Number: MFCD09835355
CAS Number: 38186-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR