tert-Butyl-4-(pyrimidin-2-yloxy)piperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-(pyrimidin-2-yloxy)piperidine-1-carboxylate

074495-5G
102536-710EA 830.77 USD
102536-710
tert-Butyl-4-(pyrimidin-2-yloxy)piperidine-1-carboxylate ≥95%
tert-Butyl-4-(pyrimidin-2-yloxy)piperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₁N₃O₃
MW: 279.3400 g/mol
MDL Number: MFCD09028557
CAS Number: 412293-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR