1-(3-Fluoropyridin-4-yl)ethanone ≥95%

Supplier: Matrix Scientific

074494-1G
102536-706EA 1596.92 USD
102536-706
1-(3-Fluoropyridin-4-yl)ethanone ≥95%
1-(3-Fluoropyridin-4-yl)ethanone

Hazard: Irritant

Formula: C₇H₆FNO
MW: 139.1300 g/mol
MDL Number: MFCD07375077
CAS Number: 87674-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR