Ethyl-2-(pyridin-3-yloxy)acetate ≥95%

Supplier: Matrix Scientific

Synonyms: (Pyridin-3-yloxy)acetic acid ethyl ester

074490-5G
102536-688EA 138.46 USD
102536-688
Ethyl-2-(pyridin-3-yloxy)acetate ≥95%
Ethyl-2-(pyridin-3-yloxy)acetate

Hazard: Irritant

Formula: C₉H₁₁NO₃
MW: 181.19
MDL Number: MFCD00093962
CAS Number: 18342-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR