4-Iodo-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

074489-5G
102536-684EA 1841.54 USD
102536-684
4-Iodo-2-methoxypyridine ≥95%
4-Iodo-2-methoxypyridine

Hazard: Irritant

Formula: C₆H₆INO
MW: 235.0224 g/mol
MDL Number: MFCD08277272
CAS Number: 98197-72-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR