4-Nitro-o-anisidine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methoxy-4-nitroaniline2-Amino-5-nitroanisole

074488-100G
102536-680EA 155.38 USD
102536-680
4-Nitro-o-anisidine ≥95%
4-Nitro-o-anisidine

Hazard: Irritant

Formula: C₇H₈N₂O₃
MW: 168.15 g/mol
MDL Number: MFCD00007363
CAS Number: 97-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR