Ethyl-7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate

074485-5G
102536-668EA 921.54 USD
102536-668
Ethyl-7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate ≥95%
Ethyl-7-chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₂ClFN₂O₃
MW: 310.7081 g/mol
MDL Number: MFCD08458313
CAS Number: 96568-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR