2,5-Xylenol ≥95%

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethylphenol2-Hydroxy-4-xylene2-Hydroxy-p-xylene

074484-100G
102536-664EA 83.08 USD
102536-664
2,5-Xylenol ≥95%
2,5-Xylenol

Hazard: Irritant

Formula: C₈H₁₀O
MW: 122.17 g/mol
MDL Number: MFCD00002237
CAS Number: 95-87-4
UN: 2261
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR