5-Aminoisoquinolin-1(2H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Aminoisoquinolin-1-ol

074475-1G
102536-622EA 640 USD
102536-622
5-Aminoisoquinolin-1(2H)-one ≥95%
5-Aminoisoquinolin-1(2H)-one

Hazard: Irritant

Formula: C₉H₈N₂O
MW: 160.1700 g/mol
MDL Number: MFCD06198896
CAS Number: 93117-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR