2,4-Dimethoxybenzoic acid ≥95%

Supplier: Matrix Scientific

074469-100G
102536-592EA 67.69 USD
102536-592
2,4-Dimethoxybenzoic acid ≥95%
2,4-Dimethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₁₀O₄
MW: 182.18 g/mol
Melting Pt: 107…111 °C
MDL Number: MFCD00002434
CAS Number: 91-52-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR