5-Chloro-2-(trifluoromethyl)benzaldehyde ≥95%

Supplier: Matrix Scientific

074464-5G
102536-566EA 246.15 USD
102536-566
5-Chloro-2-(trifluoromethyl)benzaldehyde ≥95%
5-Chloro-2-(trifluoromethyl)benzaldehyde

Hazard: Irritant

Formula: C₈H₄ClF₃O
MDL Number: MFCD03412204
CAS Number: 90381-07-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR