4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid ≥95%

Supplier: Matrix Scientific

074462-100G
102536-558EA 346.15 USD
102536-558
4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid ≥95%
4-Amino-5-hydroxynaphthalene-2,7-disulphonic acid

Hazard: Irritant

Formula: C₁₀H₉NO₇S₂
CAS Number: 90-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR