Methyl-4-aminothiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-aminothiophene-2-carboxylate

074456-1G
102536-530EA 307.69 USD
102536-530
Methyl-4-aminothiophene-2-carboxylate ≥95%
Methyl-4-aminothiophene-2-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₂S
MW: 157.1900 g/mol
MDL Number: MFCD07368326
CAS Number: 89499-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR