tert-Butyl (3-oxocyclohexyl)carbamate ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(Boc-amino)cyclohexanone

074452-5G
102536-510EA 206.15 USD
102536-510
tert-Butyl (3-oxocyclohexyl)carbamate ≥95%
tert-Butyl (3-oxocyclohexyl)carbamate

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MW: 213.27 g/mol
MDL Number: MFCD03790905
CAS Number: 885280-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR