1-Bromo-3,5-difluoro-2-nitrobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-4,6-difluoronitrobenzene

074450-5G
102536-500EA 504.62 USD
102536-500
1-Bromo-3,5-difluoro-2-nitrobenzene ≥95%
1-Bromo-3,5-difluoro-2-nitrobenzene

Hazard: Irritant

Formula: C₆H₂BrF₂NO₂
MW: 237.9900 g/mol
MDL Number: MFCD04112501
CAS Number: 884494-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR