2-(1H-Imidazol-5-yl)ethanol ≥95%
Supplier: Matrix Scientific
074442-250MG
074442-5G
102536-458EA
461.54
USD
102536-458
102536-454
2-(1H-Imidazol-5-yl)ethanol ≥95%
2-(1H-Imidazol-5-yl)ethanol
Hazard: Irritant
Formula:
C₅H₈N₂O MW: 112.1300 g/mol |
MDL Number:
MFCD00195671 CAS Number: 872-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...