4,7-Dichloroquinoline ≥95%

Supplier: Matrix Scientific

074441-10G
102536-450EA 27.6 USD
102536-450
4,7-Dichloroquinoline ≥95%
4,7-Dichloroquinoline

Hazard: Irritant

Formula: C₉H₅Cl₂N
MW: 198.05 g/mol
MDL Number: MFCD00006774
CAS Number: 86-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR