5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

074439-5G
102536-442EA 553.85 USD
102536-442
5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid ≥95%
5-Bromo-1-methyl-1H-pyrrole-2-carboxylic acid

Hazard: Irritant

Formula: C₆H₆BrNO₂
MW: 204.0213 g/mol
MDL Number: MFCD16659627
CAS Number: 865186-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR