3-Amino-4-fluorobenzonitrile ≥95%

Supplier: Matrix Scientific

074435-1G
102536-416EA 18.46 USD
102536-416
3-Amino-4-fluorobenzonitrile ≥95%
3-Amino-4-fluorobenzonitrile

Hazard: Toxic

Formula: C₇H₅FN₂
MW: 136.13 g/mol
MDL Number: MFCD07787428
CAS Number: 859855-53-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR