1-Boc-3-bromo-4-oxopyrrolidine ≥95%

Supplier: Matrix Scientific

074432-1G
102536-402EA 1383.08 USD
102536-402
1-Boc-3-bromo-4-oxopyrrolidine ≥95%
1-Boc-3-bromo-4-oxopyrrolidine

Hazard: Irritant

Formula: C₉H₁₄BrNO₃
CAS Number: 885278-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR