4-Chloro-5-nitropyridin-2(1H)-one ≥95%
Supplier: Matrix Scientific
Synonyms:
4-Chloro-2-hydroxy-5-nitropyridine
074430-5G
102536-392EA
592.31
USD
102536-392
4-Chloro-5-nitropyridin-2(1H)-one ≥95%
4-Chloro-5-nitropyridin-2(1H)-one
Hazard: Irritant
Formula:
C₅H₃ClN₂O₃ MW: 174.5400 g/mol |
MDL Number:
MFCD11046303 CAS Number: 850663-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...