1-Bromo-3-fluoro-5-iodobenzene ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Fluoro-5-iodobromobenzene

074427-5G
102536-372EA 769.23 USD
102536-372
1-Bromo-3-fluoro-5-iodobenzene ≥95%
1-Bromo-3-fluoro-5-iodobenzene

Hazard: Irritant

Formula: C₆H₃BrFI
MW: 300.9 g/mol
MDL Number: MFCD06657969
CAS Number: 845866-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR