1,3,7,8-Tetramethylxanthine ≥95%
Supplier: Matrix Scientific
074420-1G
102536-334EA
427.69
USD
102536-334
1,3,7,8-Tetramethylxanthine ≥95%
1,3,7,8-Tetramethylxanthine
Hazard: Irritant
Formula:
C₉H₁₂N₄O₂ |
MDL Number:
MFCD01741437 CAS Number: 832-66-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...