2-Bromo-4-methylbenzaldehyde ≥95%

Supplier: Matrix Scientific

074415-100G
102551-896EA 329.23 USD
102551-896
2-Bromo-4-methylbenzaldehyde ≥95%
2-Bromo-4-methylbenzaldehyde

Hazard: Irritant

Formula: C₈H₇BrO
MDL Number: MFCD04039890
CAS Number: 824-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR