2-(1-Naphthalene)ethanol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(1-Naphthyl)ethanol2-(Naphthalen-1-yl)ethanol

074400-100G
102536-234EA 150.77 USD
102536-234
2-(1-Naphthalene)ethanol ≥95%
2-(1-Naphthalene)ethanol

Hazard: Irritant

Formula: C₁₂H₁₂O
MW: 172.23 g/mol
MDL Number: MFCD00004050
CAS Number: 773-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR