5-Methoxyoxindole ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Methoxy-2-indolinone5-Methoxyindolin-2-one

074397-5G
102536-218EA 1383.08 USD
102536-218
5-Methoxyoxindole ≥95%
5-Methoxyoxindole

Hazard: Irritant

Formula: C₉H₉NO₂
MW: 163.1733 g/mol
MDL Number: MFCD01860251
CAS Number: 7699-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR