3-Cyclopentene-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: cyclopent-3-ene-1-carboxylic acidCyclopent-3-enecarboxylic acid

074395-5G
102536-208EA 23.08 USD
102536-208
3-Cyclopentene-1-carboxylic acid ≥95%
3-Cyclopentene-1-carboxylic acid

Hazard: Irritant

Formula: C₆H₈O₂
MW: 112.13 g/mol
MDL Number: MFCD00101764
CAS Number: 7686-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR