4-Bromobenzothiazole ≥95%

Supplier: Matrix Scientific

074394-1G
102536-202EA 243.08 USD
102536-202
4-Bromobenzothiazole ≥95%
4-Bromobenzothiazole

Hazard: Irritant

Formula: C₇H₄BrNS
MW: 214.0800 g/mol
MDL Number: MFCD11858590
CAS Number: 767-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR