2-Cyano-5-pyridineboronic acid pinacol ester ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyanopyridine-5-boronic acid pinacol ester2-(6-Cyano-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)picolinonitrile2-Cyano-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine6-Cyano-3-pyridylboronic acid Pinacol ester

074389-5G
102536-174EA 300 USD
102536-174
2-Cyano-5-pyridineboronic acid pinacol ester ≥95%
2-Cyano-5-pyridineboronic acid pinacol ester

Hazard: Irritant

Formula: C₁₂H₁₅BN₂O₂
MW: 230.07 g/mol
MDL Number: MFCD06657825
CAS Number: 741709-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR