2-Amino-1-(2-fluorophenyl)ethanone ≥95%

Supplier: Matrix Scientific

074387-5G
102536-164EA 1535.38 USD
102536-164
2-Amino-1-(2-fluorophenyl)ethanone ≥95%
2-Amino-1-(2-fluorophenyl)ethanone

Hazard: Irritant

Formula: C₈H₈FNO
MDL Number: MFCD08460113
CAS Number: 736887-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR